missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2',3'-O-Isopropylideneadenosine, 98%

Product Code. 10081190 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10081190 5 g Glass bottle 5g
10295400 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10081190 Supplier Thermo Scientific Chemicals Supplier No. 122720050
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 362-75-4 | C13H17N5O4 | 307.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 362-75-4
Molecular Formula C13H17N5O4
Molecular Weight (g/mol) 307.31
MDL Number MFCD00005756
InChI Key LCCLUOXEZAHUNS-AUWRGFAENA-N
Synonym 2',3'-o-isopropylideneadenosine, 3ar,4r,6r,6ar-6-6-amino-9h-purin-9-yl-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, 2',3'-isopropylideneadenosine, 3ar,4r,6r,6ar-6-6-amino-9h-purin-9-yl-2,2-dimethyl-tetrahydro-2h-furo 3,4-d 1,3 dioxol-4-yl methanol, 2 ,3-o-isopropylideneadenosine, 1r,2r,4r,5r-4-6-aminopurin-9-yl-7,7-dimethyl-3,6,8-trioxabicyclo 3.3.0 oc t-2-yl methan-1-ol, 3ar,4r,6r,6ar-4-6-aminopurin-9-yl-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro 3,4-d 1,3 dioxol-6-yl methanol, 3ar,4r,6r,6ar-6-6-aminopurin-9-yl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, pubchem14185
PubChem CID 2723654
IUPAC Name [(3aR,4R,6R,6aR)-4-(6-aminopurin-9-yl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-6-yl]methanol
SMILES CC1(C)O[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O1)N1C=NC2=C(N)N=CN=C12

Specifications

Absorbance 0.75 to 0.81 (A250/A260),0.13 to 0.17 (A280/A260),0.01 (A290/A260) max
CAS Min % 97.5
CAS Max % 100.0
Melting Point 221.0°C to 222.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Spectrophotometry 96% min. (lambda max. 259nm,pH 7) molar absorptivity 15400+/-4%,on dry substance
Formula Weight 307.3
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2', 3'-O-Isopropylideneadenosine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.