missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Piperidinyl)azepane, tech. 90%, Thermo Scientific™

Product Code. 15423357
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15423357 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15423357 Supplier Thermo Scientific Alfa Aesar Supplier No. H26042.MC
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 912771-29-0
Molecular Formula C11H22N2
MDL Number MFCD08277605
Quantity 100 mg
UN Number UN2735
Sensitivity Air Sensitive
Synonym 2-piperidin-3-yl azepane, 2-3-piperidinyl hexamethyleneimine, 2-3-piperidinyl azepane, 2-piperidin-3-yl-azepane, 2-3-piperidinyl hexahydroazepine
InChI Key NILYFVLXUNTUTG-UHFFFAOYNA-N
SMILES C1CNCC(C1)C1CCCCCN1
IUPAC Name 2-piperidin-3-ylazepane
Molecular Weight (g/mol) 182.31
PubChem CID 16740519
Formula Weight 182.18
Percent Purity 90%
Grade Technical
Chemical Name or Material 2-(3-Piperidinyl)azepane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.