missing translation for 'onlineSavingsMsg'
Learn More

2,3-Pyridinedicarboxylic Acid, TRC

Product Code. 30040358
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040358 1 g 1g
30005106 10 g 10g
18061640 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040358 Supplier Toronto Research Chemicals Supplier No. TRCP9916301G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-00-9
Molecular Formula C7 H5 N O4
Molecular Weight (g/mol) 167.12
Synonym Quinolinic Acid, Pyridine-2,3-dicarboxylic Acid, Pyridin-2,3-dicarbonsaeure, NSC 13127, NSC 18836, NSC 403247;
IUPAC Name pyridine-2,3-dicarboxylic acid
SMILES OC(=O)c1cccnc1C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H5NO4/c9-6(10)4-2-1-3-8-5(4)7(11)12/h1-3H,(H,9,10)(H,11,12)
Formula Weight 167.0219
Chemical Name or Material 2,3-Pyridinedicarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.