missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Pyridyl)-1,3-thiazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10394512
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10394512 250 mg 250mg
10354562 1 g 1g
10076944 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10394512 Supplier Thermo Scientific Maybridge Supplier No. CC13701CB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 39067-29-3
Molecular Formula C9H6N2O2S
Molecular Weight (g/mol) 206.219
MDL Number MFCD00052304
InChI Key FOQFGMAZUTUELM-UHFFFAOYSA-N
Synonym 2-pyridin-3-yl thiazole-4-carboxylic acid, 2-3-pyridyl-1,3-thiazole-4-carboxylic acid, 2-pyridin-3-yl-1,3-thiazole-4-carboxylic acid, 4-thiazolecarboxylic acid, 2-3-pyridinyl, 2-pyridin-3-yl-thiazole-4-carboxylic acid, 2-3-pyridyl thiazole-4-carboxylic acid, 2-3-pyridinyl-4-thiazolecarboxylic acid, 2-pyrid-3-yl thiazole-4-carboxylic acid, 2-3-pyridinyl-1,3-thiazole-4-carboxylic acid
PubChem CID 736513
IUPAC Name 2-pyridin-3-yl-1,3-thiazole-4-carboxylic acid
SMILES C1=CC(=CN=C1)C2=NC(=CS2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 206.22
Percent Purity 97%
Chemical Name or Material 2-(3-Pyridyl)-1, 3-thiazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.