missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Pyridyl)ethan-1-ol, Thermo Scientific™

Product Code. 10272312
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10272312 250 mg 250mg
10009421 1 g 1g
10469454 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10272312 Supplier Thermo Scientific Maybridge Supplier No. KM09875CB
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 6293-56-7
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00831047
InChI Key YPWSASPSYAWQRK-UHFFFAOYSA-N
Synonym 3-pyridineethanol, 3-2-hydroxyethyl pyridine, 2-pyridin-3-yl ethanol, 2-3-pyridyl ethan-1-ol, 2-3-pyridyl-1-ethanol, 2-pyridin-3-yl ethan-1-ol, 3-pyridylethanol, 2-3-pyridyl ethanol, pyridyl-3-methylcarbinol, 3-hydroxyethyl pyridine
PubChem CID 80515
IUPAC Name 2-pyridin-3-ylethanol
SMILES C1=CC(=CN=C1)CCO

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 123.15
Percent Purity 97%
Chemical Name or Material 2-(3-Pyridyl)ethan-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.