missing translation for 'onlineSavingsMsg'
Learn More

2-{4-[3-(Dimethylamino)propoxy]phenyl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 95%, Thermo Scientific™

Product Code. 10235364
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10235364 1 g Amber Glass Bottle 1g
10586504 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10235364 Supplier Thermo Scientific Maybridge Supplier No. CC59539DA
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 627899-90-5
Molecular Formula C17H28BNO3
Molecular Weight (g/mol) 305.23
MDL Number MFCD09702395
InChI Key ONRJKPSMQGEPHV-UHFFFAOYSA-N
Synonym 2-4-3-dimethylamino propoxy phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, n,n-dimethyl-3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenoxy propan-1-amine, dimethyl 3-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenoxy propyl amine, dimethyl 3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenoxy propyl amine, 4-3-dimethylamino propoxy benzeneboronic acid, 4-3-dimethylamino propoxy phenylboronic acid, pinacol ester, 4-3-dimethylamino propoxy phenylboronic acid,pinacol ester, dimethyl-3-4-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl phenoxy propyl amine
PubChem CID 24229609
IUPAC Name N,N-dimethyl-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]propan-1-amine
SMILES CN(C)CCCOC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 305.22
Percent Purity 95%
Chemical Name or Material 2-{4-[3-(Dimethylamino)propoxy]phenyl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.