missing translation for 'onlineSavingsMsg'
Learn More

2-{4-[3-(Dimethylamino)propoxy]phenyl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 95%, Thermo Scientific™

Product Code. p-3541031
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10586504

Brand: Thermo Scientific Maybridge CC59539CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 627899-90-5
Molecular Formula C17H28BNO3
Molecular Weight (g/mol) 305.23
MDL Number MFCD09702395
InChI Key ONRJKPSMQGEPHV-UHFFFAOYSA-N
Synonym 2-4-3-dimethylamino propoxy phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, n,n-dimethyl-3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenoxy propan-1-amine, dimethyl 3-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenoxy propyl amine, dimethyl 3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenoxy propyl amine, 4-3-dimethylamino propoxy benzeneboronic acid, 4-3-dimethylamino propoxy phenylboronic acid, pinacol ester, 4-3-dimethylamino propoxy phenylboronic acid,pinacol ester, dimethyl-3-4-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl phenoxy propyl amine
PubChem CID 24229609
IUPAC Name N,N-dimethyl-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]propan-1-amine
SMILES CN(C)CCCOC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 305.22
Percent Purity 95%
Chemical Name or Material 2-{4-[3-(Dimethylamino)propoxy]phenyl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.