missing translation for 'onlineSavingsMsg'
Learn More

2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]-1,2-oxazinane, ≥97%, Thermo Scientific™

Product Code. 11360627
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11360627 250 mg Amber Glass Bottle 250mg
10307304 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11360627 Supplier Thermo Scientific Maybridge Supplier No. CC45539CB
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 859833-22-0
Molecular Formula C18H28BNO2
Molecular Weight (g/mol) 301.24
MDL Number MFCD08271932
InChI Key HJKVRMHVLINDRH-UHFFFAOYSA-N
Synonym 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperidine, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, 4-1-piperidinylmethyl benzeneboronic acid pinacol ester, piperidine,1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl, amtb120, 4-piperidinomethyl phenylboronic acid pinacol ester, 4-piperidinomethyl phenylboronic acid, pinacol ester, 4-piperidin-1-ylmethyl phenyl boronic acid pinacol ester
PubChem CID 18525779
IUPAC Name 1-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}piperidine
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(CN2CCCCC2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 301.24
Percent Purity 97%
Chemical Name or Material 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.