missing translation for 'onlineSavingsMsg'
Learn More

2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 97%

Product Code. 10662803 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10662803 1 g Glass bottle 1g
10232572 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10662803 Supplier Thermo Scientific Acros Supplier No. 367560010
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 191171-55-8 | C12H18BNO2 | 219.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 191171-55-8
Molecular Formula C12H18BNO2
Molecular Weight (g/mol) 219.09
MDL Number MFCD02179448
InChI Key ZCJRWQDZPIIYLM-UHFFFAOYSA-N
Synonym 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-aminophenylboronic acid pinacol ester, 2-aminophenylboronic acid, pinacol ester, 2-aminophenylboronicacidpinacolester, 2-aminophenylboronic acid pinacol cyclic ester, 2-aminobenzeneboronic acid pinacol ester, 2-aminophenyl boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, 2-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, benzenamine, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 2734652
IUPAC Name 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1(C)OB(OC1(C)C)C1=CC=CC=C1N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 64.0°C to 70.0°C
Color Cream
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble in water.
Formula Weight 219.09
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.