Learn More
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Melting Point | -101°C |
| Color | Colorless |
| Density | 0.7000g/mL |
| Boiling Point | 101°C to 102°C |
| Flash Point | -6°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% |
| Packaging | Glass bottle |
| Vedi altri risultati |
Sicurezza e movimentazione
- 2,4,4-Trimethyl-1-pentene
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P273-Avoid release to the environment.
- MIXTURE LIST-Contains : 2,4,4-Trimethyl-1-pentene
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.