missing translation for 'onlineSavingsMsg'
Learn More

2,4,4-Trimethyl-1-pentene, 99%

Product Code. 11489626
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11489626

Brand: Thermo Scientific Alfa Aesar B20187.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-39-1 | C8H16 | 112.216 g/mol

2,4,4-Trimethyl-1-pentene is used in the manufacturing of chemicals, adhesives, sealant chemicals, fuels and fuel additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,4-Trimethyl-1-pentene is used in the manufacturing of chemicals, adhesives, sealant chemicals, fuels and fuel additives.

Solubility
Miscible with acetone. Partially soluble in methanol, diethyl ether, benzene, chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-39-1
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00008855
InChI Key FXNDIJDIPNCZQJ-UHFFFAOYSA-N
Synonym 2,4,4-trimethyl-1-pentene, diisobutylene, pentene, 2,4,4-trimethyl, 2,4,4-trimethylpentene, 1-pentene, 2,4,4-trimethyl, 2,2,4-trimethyl-4-pentene, unii-n69l73advf, 1-methyl-1-neopentylethylene, ccris 9103, 2,4,4-trimethylpentene-1
PubChem CID 7868
IUPAC Name 2,4,4-trimethylpent-1-ene
SMILES CC(=C)CC(C)(C)C

Specifications

Melting Point -94°C
Density 0.715
Boiling Point 101°C to 102°C
Flash Point −6°C (21°F)
Odor Petrol-like
Refractive Index 1.4085
Quantity 100 g
UN Number UN2050
Beilstein 1098309
Solubility Information Miscible with acetone. Partially soluble in methanol,diethyl ether,benzene,chloroform. Immiscible with water.
Formula Weight 112.22
Percent Purity 99%
Chemical Name or Material 2,4,4-Trimethyl-1-pentene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.