missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trichlorobenzenesulfonyl chloride, 98%

Product Code. 11443987
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11443987

Brand: Thermo Scientific Alfa Aesar A19390.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15945-07-0 | C6H2Cl4O2S | 279.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15945-07-0
Molecular Formula C6H2Cl4O2S
Molecular Weight (g/mol) 279.94
MDL Number MFCD00007428
InChI Key WNVVRCKTQSCPAC-UHFFFAOYSA-N
Synonym 2,4,5-trichlorobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4,5-trichloro, unii-7gd5283xix, 2,4,5-trichlorobenzenesulphonyl chloride, 2,4,5-trichlorobenzenesulfonylchloride, pubchem5174, acmc-1c50t, 2,4,5trichlorophenylsulfonylchloride, 2,5-trichlorobenzenesulfonyl chloride, chloro 2,4,5-trichlorophenyl sulfone
PubChem CID 27575
IUPAC Name 2,4,5-trichlorobenzenesulfonyl chloride
SMILES C1=C(C(=CC(=C1Cl)Cl)Cl)S(=O)(=O)Cl

Specifications

Melting Point 67°C to 71°C
Quantity 5 g
UN Number UN3261
Beilstein 1112595
Sensitivity Moisture sensitive
Formula Weight 279.96
Percent Purity 98%
Chemical Name or Material 2,4,5-Trichlorobenzenesulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.