missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trichlorophenol-13C6, TRC

Product Code. 30453200
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30453200 5 mg 5mg
30451878 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30453200 Supplier Toronto Research Chemicals Supplier No. TRCT7741485MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4,5-Trichlorophenol-13C6
CAS 2483736-12-3
Molecular Formula 13C6 H3 Cl3 O
Quantity 5 mg
Synonym 2,4,5-Trichlorophenol-13C6, 2,4,5-Trichlorophenic Acid-13C6, Dowicide 2-13C6, NSC 2266-13C6, Preventol I-13C6, TCP-13C6
InChI Formula InChI=1S/C6H3Cl3O/c7-3-1-5(9)6(10)2-4(3)8/h1-2,10H/i1+1,2+1,3+1,4+1,5+1,6+1
SMILES O[13c]1[13cH][13c](Cl)[13c](Cl)[13cH][13c]1Cl
IUPAC Name 2,4,5-trichloro(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-ol
Molecular Weight (g/mol) 203.4
Formula Weight 201.9451
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.