missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trichloropyrimidine, 98%

Product Code. 11348866
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348866 5 g 5g
11338866 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348866 Supplier Thermo Scientific Alfa Aesar Supplier No. L19506.06
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5750-76-5 | C4HCl3N2 | 183.416 g/mol

2,4,5-Trichloropyrimidine is used in the synthesis of potent and selective anaplastic lymphoma kinase (ALK-5) inhibitors, used as an anti-tumor treatment. It is also used in the synthesis of EGFR inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,5-Trichloropyrimidine is used in the synthesis of potent and selective anaplastic lymphoma kinase (ALK-5) inhibitors, used as an anti-tumor treatment. It is also used in the synthesis of EGFR inhibitors.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5750-76-5
Molecular Formula C4HCl3N2
Molecular Weight (g/mol) 183.416
MDL Number MFCD03788200
InChI Key GIKMWFAAEIACRF-UHFFFAOYSA-N
Synonym 2,5,6-trichloropyrimidine, 2,4,5-trichloro-pyrimidine, trichloropyrimidine, pyrimidine, 2,4,5-trichloro, 2,4,5-trichlorpyrimidin, pubchem5302, zlchem 1349, pyrimidine, trichloro, acmc-209lys, 2,5-trichloropyrimidine
PubChem CID 237259
IUPAC Name 2,4,5-trichloropyrimidine
SMILES C1=C(C(=NC(=N1)Cl)Cl)Cl

Specifications

Density 1.6
Boiling Point 84°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.573
Quantity 5 g
UN Number UN1760
Beilstein 4449
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 183.43
Percent Purity 98%
Chemical Name or Material 2,4,5-Trichloropyrimidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.