missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluoro-N'-hydroxybenzenecarboximidamide, 97%, Thermo Scientific™

Catalog Number p-3075376
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
690632-34-9
C7H5F3N2O
190.125
MFCD06200878
JWJFTTGUXZRMMI-UHFFFAOYSA-N
2,4,5-trifluoro-n-hydroxy-benzamidine, 2,4,5-trifluoro-n-hydroxybenzimidamide, hydroxyimino 2,4,5-trifluorophenyl methylamine, z-2,4,5-trifluoro-n'-hydroxybenzene-1-carboximidamide
9582832
2,4,5-trifluoro-N'-hydroxybenzenecarboximidamide
C1=C(C(=CC(=C1F)F)F)C(=NO)N
This item is not returnable. View return policy
690632-34-9
C7H5F3N2O
190.125
MFCD06200878
JWJFTTGUXZRMMI-UHFFFAOYSA-N
2,4,5-trifluoro-n-hydroxy-benzamidine, 2,4,5-trifluoro-n-hydroxybenzimidamide, hydroxyimino 2,4,5-trifluorophenyl methylamine, z-2,4,5-trifluoro-n'-hydroxybenzene-1-carboximidamide
9582832
2,4,5-trifluoro-N'-hydroxybenzenecarboximidamide
C1=C(C(=CC(=C1F)F)F)C(=NO)N
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 190.12
Percent Purity 97%
Chemical Name or Material 2, 4, 5-Trifluoro-N'-hydroxybenzenecarboximidamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'