missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorobenzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 15177422 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
15177422 5 g Glass bottle 5g
15167422 1 g Glass bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 15177422

Brand: Thermo Scientific Acros 448880050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220227-21-4
Molecular Formula C6H2ClF3O2S
Molecular Weight (g/mol) 230.59
InChI Key STCZWXXRRTWRSK-UHFFFAOYSA-N
Synonym 2,4,5-trifluorobenzene-1-sulfonyl chloride, 2,4,5-trifluorobenzenesulphonyl chloride, 2,4,5-trifluorophenyl sulfonyl chloride, 2,4,5-trifluoro-benzenesulfonyl chloride, benzenesulfonyl chloride, 2,4,5-trifluoro, pubchem5179, acmc-209fqs, ksc201o4t, chloro 2,4,5-trifluorophenyl sulfone, 2,4,5-trifluorophenylsulfonyl chloride
PubChem CID 2776979
IUPAC Name 2,4,5-trifluorobenzenesulfonyl chloride
SMILES C1=C(C(=CC(=C1F)S(=O)(=O)Cl)F)F

Specifications

Density 1.6570g/mL
Flash Point >100°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula F3C6H2SO2Cl
Quantity 5 g
Specific Gravity 1.657
Formula Weight 230.59
Percent Purity 97%
Chemical Name or Material 2, 4, 5-Trifluorobenzenesulfonyl chloride
compliance-icons
Product Identifier
  • 2,4,5-Trifluorobenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402-Store in a dry place.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.