missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trihydroxypyrimidine, 98%

Product Code. 11410937
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410937 1 g 1g
11420937 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410937 Supplier Thermo Scientific Alfa Aesar Supplier No. B23320.03
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 496-76-4 | C4H4N2O3 | 128.087 g/mol

2,4,5-Trihydroxypyrimidine studies show that it can form stable base pairs with all four bases in a DNA duplex. It has been used as oxidative DNA damage biomarker in tissue engineered skin. It acts as an antagonist of uracil in the growth of Lactobacillus casei.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 496-76-4
Molecular Formula C4H4N2O3
Molecular Weight (g/mol) 128.087
MDL Number MFCD00082987
InChI Key FQXOOGHQVPKHPG-UHFFFAOYSA-N
Synonym Isobarbituric acid
PubChem CID 96994
IUPAC Name 1,3-diazinane-2,4,5-trione
SMILES C1C(=O)C(=O)NC(=O)N1

Tekniske data

Melting Point >300°C
Quantity 1 g
Beilstein 127205
Formula Weight 128.09
Percent Purity 98%
Chemical Name or Material 2,4,5-Trihydroxypyrimidine

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.