missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trimethylthiazole, 98%

Product Code. 11489346
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489346 25 g 25g
11479346 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489346 Supplier Thermo Scientific Alfa Aesar Supplier No. A19046.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 13623-11-5 | C6H9NS | 127.205 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13623-11-5
Molecular Formula C6H9NS
Molecular Weight (g/mol) 127.205
MDL Number MFCD00005332
InChI Key BAMPVSWRQZNDQC-UHFFFAOYSA-N
Synonym 2,4,5-trimethylthiazole, trimethylthiazole, trimethyl thiazole, thiazole, trimethyl, thiazole, 2,4,5-trimethyl, trimethyl-1,3-thiazole, 2,4,5-trimethyl thiazole, thiazole, 2,4,5-trimethyl-8ci 9ci, fema no. 3325, trimethyl-thiazole
PubChem CID 61653
ChEBI CHEBI:78738
IUPAC Name 2,4,5-trimethyl-1,3-thiazole
SMILES CC1=C(SC(=N1)C)C

Specifications

Density 1.043
Boiling Point 164°C to 166°C
Flash Point 64°C (147°F)
Refractive Index 1.509
Quantity 25 g
Beilstein 107148
Formula Weight 127.21
Percent Purity 98%
Chemical Name or Material 2,4,5-Trimethylthiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.