missing translation for 'onlineSavingsMsg'
Learn More

2,4,6,8-Tetramethylcyclotetrasiloxane, 99%

Product Code. 11379295
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11379295

Brand: Thermo Scientific Alfa Aesar L16642.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2370-88-9 | C4H12O4Si4 | 236.476 g/mol

Tetramethyl-cyclotetrasiloxane is a good impregnant of photoelectric material. It is used to manufacture of modified siloxane with defined hydrogen content and chain quantity. It is also used in production of silicone polymers and acts as a precursor for the deposition of polysiloxane, cyclic siloxane, silicon dioxide, oxycarbide thin films with low dielectric constant for microelectronics and semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethyl-cyclotetrasiloxane is a good impregnant of photoelectric material. It is used to manufacture of modified siloxane with defined hydrogen content and chain quantity. It is also used in production of silicone polymers and acts as a precursor for the deposition of polysiloxane, cyclic siloxane, silicon dioxide, oxycarbide thin films with low dielectric constant for microelectronics and semiconductors.

Solubility
Miscible with most organic solvents. Immiscible with water.

Notes
Incompatible with oxidizing agents, bases and acids. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2370-88-9
Molecular Formula C4H12O4Si4
Molecular Weight (g/mol) 236.476
MDL Number MFCD00039567
InChI Key WZJUBBHODHNQPW-UHFFFAOYSA-N
Synonym 2,4,6,8-tetramethylcyclotetrasiloxane, 1,3,5,7-tetramethylcyclotetrasiloxane, tetramethylcyclotetrasiloxane, tmcts, cyclotetrasiloxane, 2,4,6,8-tetramethyl, 2,4,6,8-tetramethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, 1,2,5,7-tetramethylcyclotetrasiloxane, 2,4,6,8-tetramethyl-cyclotetrasiloxan, 2,4,6,8-tetramethylcyclotetrasiloxne, 1,3,5,7-tetramethyl cyclotetrasiloxane
PubChem CID 6327421
IUPAC Name 2,4,6,8-tetramethyl-5,7-dioxa-1,3-dioxonia-2,4-disila-6,8-disilanidacycloocta-1,3-diene
SMILES C[Si-]1O[Si-]([O+]=[Si]([O+]=[Si](O1)C)C)C

Specifications

Boiling Point 134°C to 135°C
Melting Point -69°C
Quantity 100 g
UN Number UN1993
Beilstein 1787074
Flash Point 25°C (77°F)
Solubility Information Miscible with most organic solvents. Immiscible with water.
Formula Weight 240.51
Percent Purity 99%
Refractive Index 1.387
Sensitivity Moisture sensitive
Density 0.991
Chemical Name or Material 2,4,6,8-Tetramethylcyclotetrasiloxane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.