missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Collidine, 99%

Product Code. 10412935 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Unit Size:
1 L
100mL
2.5 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10412935 1 L 1 L
10790661 100 mL 100mL
10644642 500 mL 500mL
10564684 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10412935 Supplier Thermo Scientific Acros Supplier No. 161950010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 108-75-8 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-75-8
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00006338
InChI Key BWZVCCNYKMEVEX-UHFFFAOYSA-N
Synonym 2,4,6-collidine, gamma-collidine, s-collidine, pyridine, 2,4,6-trimethyl, collidine, sym-collidine, 2,4,6-kollidin, unii-7ie4bk5j5v, 2,4,6-trimethyl-pyridine, .gamma.-collidine
PubChem CID 7953
IUPAC Name 2,4,6-trimethylpyridine
SMILES CC1=CC(C)=NC(C)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -43.0°C
Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 171.0°C to 172.0°C
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H2N)(CH3)3
Refractive Index 1.4995 to 1.5015
Quantity 1 L
Beilstein 20, 250
Fieser 01,155; 13,302
Merck Index 15, 9890
Specific Gravity 0.91
Solubility Information Solubility in water: 35g/L (20°C). Other solubilities: soluble in diluted acids,soluble in alcohol,chloroform,benzene and,toluene,miscible with ether
Formula Weight 121.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 4, 6-Collidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.