missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tri(2-pyridyl)-s-triazine, 99%

Product Code. 10082120 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10082120 5 g Glass bottle 5g
10686134 1 g Glass bottle 1g
10462875 25 g Glass bottle 25g
13428408 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10082120 Supplier Thermo Scientific Acros Supplier No. 168070050
Spotlight Offers
Use promo code 28038
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3682-35-7 | C18H12N6 | 312.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3682-35-7
Molecular Formula C18H12N6
Molecular Weight (g/mol) 312.34
MDL Number MFCD00006045
InChI Key KMVWNDHKTPHDMT-UHFFFAOYSA-N
Synonym 2,4,6-tripyridyl-s-triazine, 2,4,6-tris 2-pyridyl-s-triazine, 2,4,6-tri 2-pyridyl-1,3,5-triazine, terpyridyl-s-triazine, 2,4,6-tri 2-pyridyl-s-triazine, tptz iron reagent, 1,3,5-triazine, 2,4,6-tri-2-pyridinyl, tri-2-pyridyl-s-triazine, s-triazine, tri-2-pyridyl, 2,4,6-tris 2-pyridyl-1,3,5-triazine
PubChem CID 77258
SMILES C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 248.0°C to 252.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Merck Index 15,9926
IUPAC Name 2,4,6-tripyridin-2-yl-1,3,5-triazine
Formula Weight 312.32
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2, 4, 6-Tri(2-pyridyl)-s-triazine
Show More Show Less
compliance-icons
Product Identifier
  • 2,4,6-Tri(2-pyridyl)-s-triazine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 2,4,6-Tri(2-pyridyl)-s-triazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.