missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tri(2-pyridyl)-s-triazine, 99%

Product Code. 13428408 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
1g
25g
5g
This item is not returnable. View return policy

Product Code. 13428408

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3682-35-7 | C18H12N6 | 312.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3682-35-7
Molecular Formula C18H12N6
Molecular Weight (g/mol) 312.34
MDL Number MFCD00006045
InChI Key KMVWNDHKTPHDMT-UHFFFAOYSA-N
Synonym 2,4,6-tripyridyl-s-triazine, 2,4,6-tris 2-pyridyl-s-triazine, 2,4,6-tri 2-pyridyl-1,3,5-triazine, terpyridyl-s-triazine, 2,4,6-tri 2-pyridyl-s-triazine, tptz iron reagent, 1,3,5-triazine, 2,4,6-tri-2-pyridinyl, tri-2-pyridyl-s-triazine, s-triazine, tri-2-pyridyl, 2,4,6-tris 2-pyridyl-1,3,5-triazine
PubChem CID 77258
SMILES C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 248.0°C to 252.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 100 g
Merck Index 15,9926
IUPAC Name tris(pyridin-2-yl)-1,3,5-triazine
Formula Weight 312.32
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2, 4, 6-Tri(2-pyridyl)-s-triazine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.