missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromoaniline, TRC

Product Code. 30454050
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30454050 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454050 Supplier Toronto Research Chemicals Supplier No. TRCT771800500MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4,6-Tribromoaniline
CAS 147-82-0
Molecular Formula C6 H4 Br3 N
Quantity 500 mg
Synonym Benzenamine','2,4,6-tribromo- (9CI','ACI), 2,4,6-Tribromobenzenamine (ACI), Aniline','2,4,6-tribromo- (6CI','7CI','8CI), 2,4,6-Tribromo-phenylamine, 2,4,6-Tribromoaniline, NSC 2216, sym-Tribromoaniline
InChI Formula InChI=1S/C6H4Br3N/c7-3-1-4(8)6(10)5(9)2-3/h1-2H,10H2
SMILES Nc1c(Br)cc(Br)cc1Br
IUPAC Name 2,4,6-tribromoaniline
Molecular Weight (g/mol) 329.81
Formula Weight 326.7894
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.