missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromopyrimidine, Thermo Scientific™

Product Code. 10745052
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10745052 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10745052 Supplier Thermo Scientific Maybridge Supplier No. MAY00246DA
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 36847-11-7
Packaging Amber Glass Bottle
Molecular Formula C4HBr3N2
MDL Number MFCD00233942
Quantity 1 g
Synonym pyrimidine, 2,4,6-tribromo, pubchem6896, acmc-1ae09, 2,4,6-tribromopyrimidine
InChI Key AHEYFWKLKMOHCI-UHFFFAOYSA-N
SMILES BrC1=CC(Br)=NC(Br)=N1
IUPAC Name 2,4,6-tribromopyrimidine
Molecular Weight (g/mol) 316.78
PubChem CID 258580
Formula Weight 316.78
Percent Purity 95%
Chemical Name or Material 2, 4, 6-Tribromopyrimidine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.