missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichloro-5-methylpyrimidine, 98%

Product Code. 15441268
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15441268 1 g 1g
15461268 250 mg 250mg
15451268 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15441268 Supplier Thermo Scientific Alfa Aesar Supplier No. H55115.03
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1780-36-5 | C5H3Cl3N2 | 197.443 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1780-36-5
Molecular Formula C5H3Cl3N2
Molecular Weight (g/mol) 197.443
MDL Number MFCD00233525
InChI Key VTSWSQGDJQFXHB-UHFFFAOYSA-N
Synonym 5-methyl-2,4,6-trichloropyrimidine, pyrimidine, 2,4,6-trichloro-5-methyl, 2,4,6-trichlor-5-methylpyrimidin, pubchem7341, acmc-20a0tr, 2,4,6-trichloro-5, ksc179k8l, 2,6-trichloro-5-methylpyrimidine, methyl 2,4,6-trichloropyrimidine, 2,4,6-trichoro-5-methylpyrimidine
PubChem CID 232789
IUPAC Name 2,4,6-trichloro-5-methylpyrimidine
SMILES CC1=C(N=C(N=C1Cl)Cl)Cl

Specifications

Melting Point 65°C to 69°C
Quantity 1 g
UN Number UN1759
Formula Weight 197.45
Percent Purity 98%
Chemical Name or Material 2,4,6-Trichloro-5-methylpyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.