missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichloropyrimidine, 98%

Product Code. 11445243
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445243 100 g 100g
11425243 5 g 5g
11435243 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11445243 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' A12842.22
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3764-01-0 | C4HCl3N2 | 183.416 g/mol

It is applied In selective Suzuki coupling with arylboronic acids has been reported to give 6-arylpyrimidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied In selective Suzuki coupling with arylboronic acids has been reported to give 6-arylpyrimidines.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3764-01-0
Molecular Formula C4HCl3N2
Molecular Weight (g/mol) 183.416
MDL Number MFCD00006063
InChI Key DPVIABCMTHHTGB-UHFFFAOYSA-N
Synonym pyrimidine, 2,4,6-trichloro, 2,4,6-trichloro pyrimidine, 2,4,6-trichloro-pyrimidine, ccris 7597, 2,4,6-trichlorpyrimidin, pubchem9310, acmc-1agpv, 2,6-trichloropyrimidine, 2.4.6-trichloropyrimidin, dsstox_cid_29143
PubChem CID 77378
IUPAC Name 2,4,6-trichloropyrimidine
SMILES C1=C(N=C(N=C1Cl)Cl)Cl

Specifications

Melting Point 21°C to 25°C
Density 1.595
Boiling Point 213°C to 215°C
Flash Point >110°C (230°F)
Odor Irritating
Refractive Index 1.57
Quantity 100 g
UN Number UN1759
Beilstein 118284
Solubility Information Insoluble in water.
Formula Weight 183.43
Percent Purity 98%
Chemical Name or Material 2,4,6-Trichloropyrimidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.