missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichloropyrimidine, 99%

Product Code. 10416780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10416780 25 g Glass bottle 25g
10501581 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10416780 Supplier Thermo Scientific Acros Supplier No. 139490250
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 3764-01-0 | C4HCl3N2 | 183.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3764-01-0
Molecular Formula C4HCl3N2
Molecular Weight (g/mol) 183.42
MDL Number MFCD00006063
InChI Key DPVIABCMTHHTGB-UHFFFAOYSA-N
Synonym pyrimidine, 2,4,6-trichloro, 2,4,6-trichloro pyrimidine, 2,4,6-trichloro-pyrimidine, ccris 7597, 2,4,6-trichlorpyrimidin, pubchem9310, acmc-1agpv, 2,6-trichloropyrimidine, 2.4.6-trichloropyrimidin, dsstox_cid_29143
PubChem CID 77378
IUPAC Name 2,4,6-trichloropyrimidine
SMILES C1=C(N=C(N=C1Cl)Cl)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 23°C to 25°C
Color Colorless to Yellow
Boiling Point 210°C to 215°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5690 to 1.571
Quantity 25 g
Beilstein 23, 90
Formula Weight 183.42
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material 2, 4, 6-Trichloropyrimidine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4,6-Trichloropyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.