missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorobenzenesulfonyl chloride, 97%

Product Code. 15435507
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15435507 250 mg 250mg
15425507 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435507 Supplier Thermo Scientific Alfa Aesar Supplier No. H32746.MD
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 220239-64-5 | C6H2ClF3O2S | 230.585 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220239-64-5
Molecular Formula C6H2ClF3O2S
Molecular Weight (g/mol) 230.585
MDL Number MFCD01091015
InChI Key XINCBNCLCIQIJM-UHFFFAOYSA-N
Synonym 2,4,6-trifluorobenzene-1-sulfonyl chloride, 2,4,6-trifluorobenzenesulphonyl chloride, pubchem5181, chloro 2,4,6-trifluorophenyl sulfone, 2,4,6-trifluorobenzenesulfonylchloride, 2,4,6-trifluorophenylsulfonyl chloride, benzenesulfonylchloride, 2,4,6-trifluoro, 2,4,6-trifluorobenzenesulfonyl chloride, benzenesulfonyl chloride, 2,4,6-trifluoro-9ci
PubChem CID 2776983
IUPAC Name 2,4,6-trifluorobenzenesulfonyl chloride
SMILES C1=C(C=C(C(=C1F)S(=O)(=O)Cl)F)F

Specifications

Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 230.59
Percent Purity 97%
Chemical Name or Material 2,4,6-Trifluorobenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.