missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorobenzylamine, TRC

Product Code. 30449525
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449525 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449525 Supplier Toronto Research Chemicals Supplier No. TRCT7900651G
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4,6-Trifluorobenzylamine
CAS 214759-21-4
Molecular Formula C7H6F3N
Quantity 1 g
Synonym Benzenemethanamine','2,4,6-trifluoro-, 2,4,6-Trifluorobenzenemethanamine, (2,4,6-Trifluorophenyl)methanamine, 2,4,6-Trifluorobenzylamine
InChI Formula InChI=1S/C7H6F3N/c8-4-1-6(9)5(3-11)7(10)2-4/h1-2H,3,11H2
SMILES NCc1c(F)cc(F)cc1F
IUPAC Name (2,4,6-trifluorophenyl)methanamine
Molecular Weight (g/mol) 161.12
Formula Weight 161.0452
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.