missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluoropyridine, 97%

Product Code. 11388926
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11388926

Brand: Thermo Scientific Alfa Aesar L19541.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3512-17-2 | C5H2F3N | 133.073 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3512-17-2
Molecular Formula C5H2F3N
Molecular Weight (g/mol) 133.073
MDL Number MFCD03001161
InChI Key UZDRWXKBKVVUTE-UHFFFAOYSA-N
Synonym 2,4,6-trifluoro-pyridine, pyridine, 2,4,6-trifluoro, pubchem7794, 2,4,6 trifluoropyridine, acmc-1cp7f, 2,4,6-tris fluoranyl pyridine
PubChem CID 2783289
IUPAC Name 2,4,6-trifluoropyridine
SMILES C1=C(C=C(N=C1F)F)F

Specifications

Density 1.382
Boiling Point 102°C
Flash Point 17°C (62°F)
Refractive Index 1.413
Quantity 25 g
UN Number UN2924
Beilstein 1364338
Formula Weight 133.07
Percent Purity 97%
Chemical Name or Material 2,4,6-Trifluoropyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.