missing translation for 'onlineSavingsMsg'
Learn More

2',4',6'-Trihydroxyacetophenone hydrate, 98+%

Product Code. 11366816
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11366816 25 g 25g
11356816 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11366816 Supplier Thermo Scientific Alfa Aesar Supplier No. L04156.14
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 480-66-0 | C8H8O4 | 168.15 g/mol

2',4',6'-Trihydroxyacetophenone hydrate is a compound that enhances bile secretion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,4′,6′-Trihydroxyacetophenone hydrate is a compound that enhances bile secretion.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 480-66-0
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00002287
InChI Key XLEYFDVVXLMULC-UHFFFAOYSA-N
Synonym 1-2,4,6-trihydroxyphenyl ethanone, 2',4',6'-trihydroxyacetophenone, phloroacetophenone, phloracetophenone, acetophloroglucine, 2,4,6-trihydroxyacetophenone, acetylphloroglucinol, ethanone, 1-2,4,6-trihydroxyphenyl, 2-acetylphloroglucinol, 1-2,4,6-trihydroxyphenyl-ethanone
PubChem CID 68073
ChEBI CHEBI:64344
IUPAC Name 1-(2,4,6-trihydroxyphenyl)ethanone
SMILES CC(=O)C1=C(O)C=C(O)C=C1O

Specifications

Melting Point 218°C to 223°C
Quantity 25 g
Beilstein 1911197
Solubility Information Soluble in water.
Formula Weight 168.15 (Anhydrous)
Percent Purity ≥98%
Chemical Name or Material 2',4',6'-Trihydroxyacetophenone hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.