missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triisopropylbenzenesulfonyl Chloride, TRC

Product Code. 30452856
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity unitSize
30452856 100 g 100g
1 options
This item is not returnable. View return policy

Product Code. 30452856

Brand: Toronto Research Chemicals TRCT795575100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4,6-Triisopropylbenzenesulfonyl Chloride
CAS 6553-96-4
Molecular Formula C15H23ClO2S
Quantity 100 g
Synonym 2,4,6-Tris(isopropyl)benzenesulfonyl Chloride, 2,4,6-Triisopropylbenzene-1-sulfonyl Chloride, 2,4,6-Triisopropylphenylsulfonyl Chloride, 2,4,6-Tris(isopropyl)benzenesulfonyl Chloride, NSC 102803, Tripsyl Chloride
InChI Formula InChI=1S/C15H23ClO2S/c1-9(2)12-7-13(10(3)4)15(19(16,17)18)14(8-12)11(5)6/h7-11H,1-6H3
SMILES CC(C)c1cc(C(C)C)c(c(c1)C(C)C)S(=O)(=O)Cl
IUPAC Name 2,4,6-tri(propan-2-yl)benzenesulfonyl chloride
Molecular Weight (g/mol) 302.86
Formula Weight 302.1107
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.