missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethoxybenzeneboronic acid, 98%

Product Code. 11399815
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399815 1 g 1g
11309825 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399815 Supplier Thermo Scientific Alfa Aesar Supplier No. L19837.03
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 135159-25-0 | C9H13BO5 | 212.01 g/mol

2,4,6-Trimethoxybenzeneboronic acid is used as an intermediate in organic syntheses and in Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethoxybenzeneboronic acid is used as an intermediate in organic syntheses and in Suzuki reaction.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135159-25-0
Molecular Formula C9H13BO5
Molecular Weight (g/mol) 212.01
MDL Number MFCD01114642
InChI Key PKLRXZVPEQJTTJ-UHFFFAOYSA-N
Synonym 2,4,6-trimethoxybenzeneboronic acid, 2,4,6-trimethoxyphenyl boronic acid, boronic acid,b-2,4,6-trimethoxyphenyl, 1,3,5-trimethoxybenzene-2-boronic ester, pubchem9566, 1,3,5-trimethoxybenzene-2-bronic ester, acmc-1cirz, ablock ab-12-9177, 2,4,6-trimethoxybenzeneboronicacid
PubChem CID 4197996
IUPAC Name (2,4,6-trimethoxyphenyl)boronic acid
SMILES COC1=CC(OC)=C(B(O)O)C(OC)=C1

Specifications

Melting Point 96°C to 100°C
Quantity 1 g
Beilstein 4315581
Solubility Information Slightly soluble in water.
Formula Weight 212.01
Percent Purity 98%
Chemical Name or Material 2,4,6-Trimethoxybenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.