missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-trimethyl-1,3,5-triazinane trihydrate, 95%, BLD Pharm™

Product Code. 31105013
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31105013 100 g
31103755 5 g
31099805 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31105013 Supplier BLD Pharm Supplier No. BD133216100g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 58052-80-5
Molecular Formula C6H21N3O3
MDL Number MFCD00149559
Quantity 100 g
SMILES CC1NC(C)NC(C)N1.[H]O[H].[H]O[H].[H]O[H]
Molecular Weight (g/mol) 183.25
Percent Purity 95%
Chemical Name or Material 2,4,6-trimethyl-1,3,5-triazinane trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.