missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylbenzyl Chloride, TRC

Product Code. 30454308
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30454308 10 g 10g
30451593 5 g 5g
30449000 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30454308 Supplier Toronto Research Chemicals Supplier No. TRCT79595510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1585-16-6
Molecular Formula C10 H13 Cl
Molecular Weight (g/mol) 168.66
IUPAC Name 2-(chloromethyl)-1,3,5-trimethylbenzene
SMILES Cc1cc(C)c(CCl)c(C)c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C10H13Cl/c1-7-4-8(2)10(6-11)9(3)5-7/h4-5H,6H2,1-3H3
Formula Weight 168.0706
Chemical Name or Material 2,4,6-Trimethylbenzyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.