missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylphenyl isocyanate, 98%

Product Code. 11353266
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353266 1 g 1g
11363266 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11353266 Supplier Thermo Scientific Alfa Aesar Supplier No. L12611.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2958-62-5 | C10H11NO | 161.204 g/mol

2,4,6-Trimethylphenyl isocyanate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylphenyl isocyanate is used as an organic chemical synthesis intermediate.

Solubility
It hydrolyzes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols, amines, bases and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2958-62-5
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.204
MDL Number MFCD00019909
InChI Key HJHZRZFONUPQAA-UHFFFAOYSA-N
Synonym 2,4,6-trimethylphenyl isocyanate, mesityl isocyanate, 2,4,6-trimethylphenylisocyanate, acmc-1ckns, 2,4,6-trimethylbenzenisocyanate, 2-isocyanato-1,3,5-trimethyl-benzene, #
PubChem CID 595239
IUPAC Name 2-isocyanato-1,3,5-trimethylbenzene
SMILES CC1=CC(=C(C(=C1)C)N=C=O)C

Specifications

Melting Point 43°C to 46°C
Boiling Point 218°C to 220°C
Flash Point 104°C (219°F)
Odor Irritating
Quantity 1 g
UN Number UN2811
Beilstein 2085601
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes in water.
Formula Weight 161.21
Percent Purity 98%
Chemical Name or Material 2,4,6-Trimethylphenyl isocyanate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4,6-Trimethylphenyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H330-Fatal if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P320-Specific treatment is urgent (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.