missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triphenylpyrylium tetrafluoroborate, 97%

Product Code. 11463747
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463747 100 g 100g
11443747 5 g 5g
11453747 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463747 Supplier Thermo Scientific Alfa Aesar Supplier No. A18217.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 448-61-3 | C23H17BF4O | 396.19 g/mol

2,4,6-Triphenylpyrylium tetrafluoroborate is used as a sensitizer to carry out the photo oxidation of cathecol. It is also used in the preparation of three N-alkylpyridinium photosensitizers by reacting with (1R,2S)-(-)-norephedrine, (S)-(+)-2-(aminomethyl)pyrrolidine and (R)-(-)-1-cyclohexylethylamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Triphenylpyrylium tetrafluoroborate is used as a sensitizer to carry out the photo oxidation of cathecol. It is also used in the preparation of three N-alkylpyridinium photosensitizers by reacting with (1R,2S)-(-)-norephedrine, (S)-(+)-2-(aminomethyl)pyrrolidine and (R)-(-)-1-cyclohexylethylamine.

Solubility
Insoluble in water, ethyl alcohol, diethyl ether. Soluble in trifluroacetic acid.

Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 448-61-3
Molecular Formula C23H17BF4O
Molecular Weight (g/mol) 396.19
MDL Number MFCD00012001
InChI Key VQYPWMWEJGDSTF-UHFFFAOYSA-N
Synonym 2,4,6-triphenylpyrylium tetrafluoroborate, 2,4,6-triphenyl-1??-pyran-1-ylium tetrafluoroborate, 2,4,6-triphenylpyryliumtetrafluoroborate, pyrylium, 2,4,6-triphenyl-, tetrafluoroborate 1-, 2,4,6-triphenyl-pyrylium tetrafluoroborate
PubChem CID 9930615
IUPAC Name 2,4,6-triphenylpyrylium;tetrafluoroborate
SMILES F[B-](F)(F)F.C1=CC=C(C=C1)C1=CC(=[O+]C(=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 248°C to 254°C
Quantity 100 g
Beilstein 3586533
Sensitivity Moisture sensitive
Solubility Information Insoluble in water,ethyl alcohol,diethyl ether. Soluble in trifluroacetic acid.
Formula Weight 396.2
Percent Purity 97%
Chemical Name or Material 2,4,6-Triphenylpyrylium tetrafluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.