missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tris(allyloxy)-1,3,5-triazine, 98%, stab. with 100ppm hydroquinone

Product Code. 15410847
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15410847 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15410847 Supplier Thermo Scientific Alfa Aesar Supplier No. L16274.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 101-37-1 | C12H15N3O3 | 249.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 101-37-1
Melting Point 25°C to 28°C
Boiling Point 149°C to 150°C (4 mmHg)
Flash Point 166°C (331°F)
Molecular Formula C12H15N3O3
MDL Number MFCD00006049
Quantity 50 g
UN Number UN3077
Synonym triallyl cyanurate, 2,4,6-triallyloxy-1,3,5-triazine, triallyl cyanaurate, activator oc, rhenofit tac, cyanuric acid triallyl ester, perkalink 300, tripropargyl cyanurate, perkalink 300-50d, 1,3,5-triazine, 2,4,6-tris 2-propenyloxy
InChI Key BJELTSYBAHKXRW-UHFFFAOYSA-N
SMILES C=CCOC1=NC(=NC(=N1)OCC=C)OCC=C
IUPAC Name 2,4,6-tris(prop-2-enoxy)-1,3,5-triazine
Molecular Weight (g/mol) 249.27
PubChem CID 7555
Formula Weight 249.27
Percent Purity 98%
Chemical Name or Material 2,4,6-Tris(allyloxy)-1,3,5-triazine, stabilized with 100ppm hydroquinone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.