missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine, Mass Spec Std.

Product Code. p-7046944
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11343816 1 g 1g
11353816 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343816 Supplier Thermo Scientific Alfa Aesar Supplier No. L16883.03
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 915-76-4 | C12F21N3 | 585.119 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 915-76-4
Molecular Formula C12F21N3
Molecular Weight (g/mol) 585.119
MDL Number MFCD00042438
InChI Key KXQUYHRRCVECPV-UHFFFAOYSA-N
Synonym tris heptafluoropropyl-s-triazine, s-triazine, 2,4,6-tris heptafluoropropyl, 2,4,6-tris heptafluoropropyl-1,3,5-triazine, tris perfluoropropyl-s-triazine, 1,3,5-triazine, 2,4,6-tris heptafluoropropyl, 2,4,6-tris perfluoropropyl-1,3,5-triazine, perfluorotripropyl-s-triazine, tris heptafluoropropyl-1,3,5-triazine, s-triazine,4,6-tris heptafluoropropyl, 1,5-triazine, 2,4,6-tris heptafluoropropyl
PubChem CID 70185
IUPAC Name 2,4,6-tris(1,1,2,2,3,3,3-heptafluoropropyl)-1,3,5-triazine
SMILES C1(=NC(=NC(=N1)C(C(C(F)(F)F)(F)F)(F)F)C(C(C(F)(F)F)(F)F)(F)F)C(C(C(F)(F)F)(F)F)(F)F

Specifications

Density 1.716
Boiling Point 165°C
Refractive Index 1.311
Quantity 1 g
Beilstein 381010
Formula Weight 585.12
Chemical Name or Material 2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine, Mass Spec Standard

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.