missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol, 96%

Product Code. 11416966
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416966 1 g 1g
11426966 5 g 5g
11436966 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11416966 Supplier Thermo Scientific Alfa Aesar Supplier No. B24048.03
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 722-92-9 | C9H7F6NO | 259.151 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 722-92-9
Molecular Formula C9H7F6NO
Molecular Weight (g/mol) 259.151
MDL Number MFCD00039258
InChI Key TZEJXCIGVMTMDY-UHFFFAOYSA-N
Synonym 2-4-aminophenyl-1,1,1,3,3,3-hexafluoropropan-2-ol, 4-hexafluoro-2-hydroxyisopropyl aniline, 2-4-aminophenyl-1,1,1,3,3,3-hexafluoro-2-propanol, 2-4-aminophenyl hexafluoro-2-propanol, 2-4-amino-phenyl-1,1,1,3,3,3-hexafluoro-propan-2-ol, benzenemethanol,4-amino-a,a-bis trifluoromethyl, enamine_005408, 4-hexafluoro-2-hydroxy-2-propyl aniline, 4-hexafluoro-2-hydroxy-2-propyl-aniline, 2-4-aminophenyl hexafluoropropan-2-ol
PubChem CID 577801
IUPAC Name 2-(4-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
SMILES C1=CC(=CC=C1C(C(F)(F)F)(C(F)(F)F)O)N

Specifications

Melting Point 147°C to 152°C
Quantity 1 g
Beilstein 1473737
Formula Weight 259.15
Percent Purity 96%
Chemical Name or Material 2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.