missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Aminophenyl)ethanol, 97%

Product Code. 11358216
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11358216 5 g 5g
11368216 25 g 25g
11378216 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11358216 Supplier Thermo Scientific Alfa Aesar Supplier No. L15223.06
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 104-10-9 | C8H11NO | 137.182 g/mol

2-(4-Aminophenyl)ethanol is used as nonsymmetric monomer in the preparation of ordered [head-to-head (H-H) or tail-to-tail (T-T)] poly(amide-ester), in the synthesis of 4-aminostyrene and in functionalization of graphene nanoplatelets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-Aminophenyl)ethanol is used as nonsymmetric monomer in the preparation of ordered [head-to-head (H-H) or tail-to-tail (T-T)] poly(amide-ester), in the synthesis of 4-aminostyrene and in functionalization of graphene nanoplatelets.

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. It is sensitive to air. Incompatible with strong oxidizing agents. Store under dry inert gas. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-10-9
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00007922
InChI Key QXHDYMUPPXAMPQ-UHFFFAOYSA-N
Synonym 2-4-aminophenyl ethanol, 4-aminophenethyl alcohol, p-aminophenethyl alcohol, p-aminophenylethanol, benzeneethanol, 4-amino, 2-p-aminophenyl ethanol, 2-4-aminophenyl ethan-1-ol, 4-2-hydroxyethyl aniline, benzeneethanol, ar-amino, p-2-hydroxyethyl aniline
PubChem CID 66904
IUPAC Name 2-(4-aminophenyl)ethanol
SMILES C1=CC(=CC=C1CCO)N

Specifications

Melting Point 108°C to 110°C
Quantity 5 g
Beilstein 907205
Solubility Information Soluble in methanol.
Formula Weight 137.18
Percent Purity 97%
Chemical Name or Material 2-(4-Aminophenyl)ethanol
compliance-icons
Product Identifier
  • 2-(4-Aminophenyl)ethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-(4-Aminophenyl)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.