missing translation for 'onlineSavingsMsg'
Learn More

2,4'-Bipyridine, TRC

Product Code. 30337235
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30337235 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30337235 Supplier Toronto Research Chemicals Supplier No. TRCB59218310G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4'-Bipyridine
CAS 581-47-5
Molecular Formula C10H8N2
Quantity 10 g
Synonym 2,4'-Bipyridyl, 2,4'-Dipyridine, 2,4'-Dipyridyl, 2-(Pyridin-4-yl)pyridine, 2-Pyridin-4-ylpyridine, 4-(2-Pyridyl)pyridine
InChI Formula InChI=1S/C10H8N2/c1-2-6-12-10(3-1)9-4-7-11-8-5-9/h1-8H
SMILES c1ccc(nc1)c2ccncc2
IUPAC Name 2-pyridin-4-ylpyridine
Molecular Weight (g/mol) 156.18
Formula Weight 156.0687
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.