missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromo-2-chlorophenyl)ethyl Iodide, TRC

Product Code. 30331617
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30331617 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331617 Supplier Toronto Research Chemicals Supplier No. TRCB68224050MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(4-Bromo-2-chlorophenyl)ethyl Iodide
CAS 916516-91-1
Molecular Formula C8H7BrClI
Quantity 50 mg
Synonym 4-Bromo-2-chloro-1-(2-iodoethyl)-benzene
InChI Formula InChI=1S/C8H7BrClI/c9-7-2-1-6(3-4-11)8(10)5-7/h1-2,5H,3-4H2
SMILES Clc1cc(Br)ccc1CCI
IUPAC Name 4-bromo-2-chloro-1-(2-iodoethyl)benzene
Molecular Weight (g/mol) 345.4
Formula Weight 343.8464
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.