missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chloro-3-nitrobenzoyl)benzoic Acid, TRC

Product Code. 30351358
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30351358 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351358 Supplier Toronto Research Chemicals Supplier No. TRCC36843525G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(4-Chloro-3-nitrobenzoyl)benzoic Acid
CAS 85-54-1
Molecular Formula C14H8ClNO5
Quantity 25 g
Synonym 2-(3-Nitro-4-chlorobenzoyl)benzoic acid, 2-(4'-Chloro-3'-nitrobenzoyl)benzoic acid, 2-[(4-Chloro-3-nitrophenyl)carbonyl]benzoic acid, 3'-Nitro-4'-chlorobenzoylbenzoic acid, 4-Chloro-3-nitrobenzophenone-2'-carboxylic acid, NSC 15366
InChI Formula InChI=1S/C14H8ClNO5/c15-11-6-5-8(7-12(11)16(20)21)13(17)9-3-1-2-4-10(9)14(18)19/h1-7H,(H,18,19)
SMILES OC(=O)c1ccccc1C(=O)c2ccc(Cl)c(c2)[N+](=O)[O-]
IUPAC Name 2-(4-chloro-3-nitrobenzoyl)benzoic acid
Molecular Weight (g/mol) 305.67
Formula Weight 305.0091
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.