missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorobenzyl)pyridine, 97%, Thermo Scientific™

Product Code. 11450757
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 11450757

Brand: Thermo Scientific Alfa Aesar B23170.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 8°C
CAS 4350-41-8
Density 1.17
Boiling Point 181°C to 183°C (20 mmHg)
Quantity 25 g
Refractive Index 1.587
Molecular Formula C12H10ClN
MDL Number MFCD00006353
Beilstein 133425
Synonym 2-4-chlorobenzyl pyridine, 2-4-chlorophenyl methyl pyridine, 2-p-chlorobenzyl pyridine, 2-4'-chlorobenzyl pyridine, pyridine, 2-4-chlorophenyl methyl, 2-para-chlorobenzyl-pyridine, 2-4-chloro-benzyl-pyridine, pubchem20810, acmc-209jus, 2-p-chlorobenzylpyridine
InChI Key XSVWMIMFDMJQRL-UHFFFAOYSA-N
SMILES C1=CC=NC(=C1)CC2=CC=C(C=C2)Cl
IUPAC Name 2-[(4-chlorophenyl)methyl]pyridine
Molecular Weight (g/mol) 203.669
PubChem CID 78054
Formula Weight 203.67
Percent Purity 97%
Chemical Name or Material 2-(4-Chlorobenzyl)pyridine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.