missing translation for 'onlineSavingsMsg'
Learn More

2-[4-(chloromethyl)piperidino]pyrimidine, Thermo Scientific™

Product Code. 10785822
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10785822 250 mg Amber Glass Bottle 250mg
10677390 1 g Amber Glass Bottle 1g
10743522 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10785822 Supplier Thermo Scientific Maybridge Supplier No. CC20724CB
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 111247-62-2
Molecular Formula C10H14ClN3
Molecular Weight (g/mol) 211.693
InChI Key XEOONRMTEFSPSW-UHFFFAOYSA-N
Synonym 2-4-chloromethyl piperidin-1-yl pyrimidine, 2-4-chloromethyl piperidino pyrimidine, 2-4-chloromethyl-piperidin-1-yl-pyrimidine, 2-4-chloromethyl piperidyl pyrimidine, 2-4-chloromethylpiperidin-1-yl pyrimidine
PubChem CID 14554091
IUPAC Name 2-[4-(chloromethyl)piperidin-1-yl]pyrimidine
SMILES C1CN(CCC1CCl)C2=NC=CC=N2

Specifications

Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 211.69
Percent Purity 97%
Chemical Name or Material 2-[4-(chloromethyl)piperidino]pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.