missing translation for 'onlineSavingsMsg'
Learn More

2-(4-chlorophenoxy)-2-methylpropanenitrile, 97%, Thermo Scientific™

Product Code. 10354102
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10354102 1 g Amber Glass Bottle 1g
10531122 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10354102 Supplier Thermo Scientific Maybridge Supplier No. RDP00164DA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 24889-11-0
Molecular Formula C10H10ClNO
Molecular Weight (g/mol) 195.65
MDL Number MFCD00203862
InChI Key JENFRDUXBGWJGH-UHFFFAOYSA-N
Synonym 2-4-chlorophenoxy-2-methylpropanenitrile, 2-p-chlorophenoxy-2-methyl-propionitrile, 2-p-chloro-phenoxy-2-methyl-propionitrile, propanenitrile,2-4-chlorophenoxy-2-methyl, propanenitrile, 2-4-chlorophenoxy-2-methyl
PubChem CID 2781321
IUPAC Name 2-(4-chlorophenoxy)-2-methylpropanenitrile
SMILES CC(C)(OC1=CC=C(Cl)C=C1)C#N

Specifications

CAS Min % 97.0
Quantity 1 g
Packaging Amber Glass Bottle
Assay Percent Range 97%
Linear Formula ClC6H4OC(CH3)2CN
Formula Weight 195.65
Percent Purity 97%
Chemical Name or Material 2-(4-chlorophenoxy)-2-methylpropanenitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.