missing translation for 'onlineSavingsMsg'
Learn More

(2-(4-Chlorophenyl)-1,3-thiazol-4-yl)methanamine hydrochloride, 97%, Tech., Thermo Scientific™

Product Code. 10737885
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10737885 5 g 5g
10196802 250 mg 250mg
10055682 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10737885 Supplier Thermo Scientific Maybridge Supplier No. MO01299DE
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 690632-35-0
Molecular Formula C10H10Cl2N2S
Molecular Weight (g/mol) 261.16
MDL Number MFCD05865127
InChI Key PNBZPHKDYJHRSP-UHFFFAOYSA-N
Synonym 2-4-chlorophenyl-1,3-thiazol-4-yl methanamine hydrochloride, 2-4-chlorophenyl thiazol-4-yl methanamine hydrochloride, 2-4-chlorophenyl-1,3-thiazol-4-yl methanaminehydrochloride, bestipharma 584-709, 2-4-chloro-phenyl-thiazol-4-ylmethylamine hcl, 2-4-chlorophenyl-4-thiazolyl methanamine hydrochloride, 2-4-chlorophenyl-1,3-thiazol-4-yl methylamine, chloride, 2-4-chloro-phenyl-thiazol-4-ylmethylamine hydrochloride, 1-2-4-chlorophenyl-1,3-thiazol-4-yl methanamine hydrochloride
PubChem CID 2794737
SMILES Cl.NCC1=CSC(=N1)C1=CC=C(Cl)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
IUPAC Name [2-(4-chlorophenyl)-1,3-thiazol-4-yl]methanamine;hydrochloride
Formula Weight 261.18
Percent Purity 97%
Grade Technical
Chemical Name or Material [2-(4-Chlorophenyl)-1, 3-thiazol-4-yl]methanamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.