missing translation for 'onlineSavingsMsg'
Learn More

2-(4-chlorophenyl)-1H-indole-3-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10417252
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10417252 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10417252 Supplier Thermo Scientific Maybridge Supplier No. SEW03034CB
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 1217-83-0
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C15H10ClNO
MDL Number MFCD00047160
Quantity 250 mg
Synonym 2-4-chlorophenyl-1h-indole-3-carbaldehyde, 1h-indole-3-carboxaldehyde,2-4-chlorophenyl, 2-4-chlorophenyl-3-formyl-indole, 2-4-chlorophenyl indole-3-carbaldehyde, 2-4-chlorophenyl indole-3-carboxaldehyde
InChI Key LISKDSDZIWRRTD-UHFFFAOYSA-N
SMILES ClC1=CC=C(C=C1)C1=C(C=O)C2=CC=CC=C2N1
IUPAC Name 2-(4-chlorophenyl)-1H-indole-3-carbaldehyde
Molecular Weight (g/mol) 255.70
PubChem CID 2797336
Formula Weight 255.7
Percent Purity 97%
Chemical Name or Material 2-(4-Chlorophenyl)-1H-indole-3-carbaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.