missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenyl)ethyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 11317696
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317696 5 g 5g
11307696 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317696 Supplier Thermo Scientific Alfa Aesar Supplier No. L11307.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17608-10-5
Molecular Formula C9H8ClNS
Molecular Weight (g/mol) 197.68
MDL Number MFCD00041127
InChI Key MRJJYUJULSZFDV-UHFFFAOYSA-N
Synonym 2-4-chlorophenyl ethyl isothiocyanate, 1-chloro-4-2-isothiocyanatoethyl benzene, 2-4-chlorophenethyl isothiocyanate, 2-4-chlorophenethyl isothicyanate, acmc-1bsas, 2-4-chlorophenyl ethanisothiocyanate, 1-chloranyl-4-2-isothiocyanatoethyl benzene, 1-chloro-4-2-isothiocyanatoethyl benzene #
PubChem CID 140257
IUPAC Name 1-chloro-4-(2-isothiocyanatoethyl)benzene
SMILES C1=CC(=CC=C1CCN=C=S)Cl

Specifications

Density 1.215
Quantity 5 g
Flash Point >110°C (230°F)
Packaging Glass bottle
Refractive Index 1.5995
UN Number UN2922
Beilstein 2719060
Boiling Point 145°C (1 mmHg)
Formula Weight 197.69
Percent Purity 97%
Chemical Name or Material 2-(4-Chlorophenyl)ethyl isothiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.